COM448660511 Combi-Blocks, Inc.

Perfluoroallylfluorosulfate

  • MDL number MFCD02093345
  • Cas Number : 67641-28-5
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7281

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7281
Availability details for
Quantity
  • Availability
  • Add to Cart