COM448660588 Combi-Blocks, Inc.

3-Methoxycyclohexene

  • MDL number MFCD00013777
  • Cas Number : 2699-13-0
  • Molecular Weight : 112.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7363

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7363
Availability details for
Quantity
  • Availability
  • Add to Cart