COM448660697 Combi-Blocks, Inc.

Acetoacetic acid n-propyl ester

  • MDL number MFCD00059405
  • Cas Number : 1779-60-8
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7478

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7478
Availability details for
Quantity
  • Availability
  • Add to Cart