COM448660763 Combi-Blocks, Inc.

2-(3-(Trifluoromethyl)pyridin-2-yl)acetic acid

  • MDL number MFCD09924925
  • Cas Number : 1000521-27-6
  • Molecular Weight : 205.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7550

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7550
Availability details for
Quantity
  • Availability
  • Add to Cart