COM448660788 Combi-Blocks, Inc.

2-Isopropylmorpholine hydrochloride

  • MDL number MFCD24369144
  • Cas Number : 1432678-91-5
  • Molecular Weight : 165.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7578
Availability details for
Quantity
  • Availability
  • Add to Cart