COM448660894 Combi-Blocks, Inc.

Ethyl 2-(2-(methylthio)-6-oxo-1,6-dihydropyrimidin-5-yl)acetate

  • MDL number MFCD11977459
  • Cas Number : 29571-44-6
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7689

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7689
Availability details for
Quantity
  • Availability
  • Add to Cart