COM448661091 Combi-Blocks, Inc.

Diiododifluoromethane

  • MDL number MFCD00798131
  • Cas Number : 1184-76-5
  • Molecular Weight : 303.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7900

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7900
Availability details for
Quantity
  • Availability
  • Add to Cart