COM448661109 Combi-Blocks, Inc.

Ethyl 2-(methylsulfonyl)propanoate

  • MDL number MFCD16045025
  • Cas Number : 73017-82-0
  • Molecular Weight : 180.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7918
Availability details for
Quantity
  • Availability
  • Add to Cart