COM448661189 Combi-Blocks, Inc.

3,5-Difluoro-4-methylaniline

  • MDL number MFCD03094196
  • Cas Number : 878285-13-3
  • Molecular Weight : 143.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8006

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8006
Availability details for
Quantity
  • Availability
  • Add to Cart