COM448661209 Combi-Blocks, Inc.

3-Chloroquinolin-8-amine

  • MDL number MFCD11040215
  • Cas Number : 139399-66-9
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8027
Availability details for
Quantity
  • Availability
  • Add to Cart