COM448661276 Combi-Blocks, Inc.

Perfluoro-3,7-dimethyloctanoic acid

  • MDL number MFCD01320738
  • Cas Number : 172155-07-6
  • Molecular Weight : 514.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8100
Availability details for
Quantity
  • Availability
  • Add to Cart