COM448661312 Combi-Blocks, Inc.

(R)-2-Amino-2-(2-chlorophenyl)cyclohexanone HCl

  • MDL number MFCD13186823
  • Cas Number : 83777-68-8
  • Molecular Weight : 260.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8141
Availability details for
Quantity
  • Availability
  • Add to Cart