COM448661371 Combi-Blocks, Inc.

16-Mercapto-1-hexadecanol

  • MDL number MFCD03701687
  • Cas Number : 114896-32-1
  • Molecular Weight : 274.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8207

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8207
Availability details for
Quantity
  • Availability
  • Add to Cart