COM448661426 Combi-Blocks, Inc.

5-Methyl-3-(trifluoromethyl)-1h-pyrazole

  • MDL number MFCD00156058
  • Cas Number : 10010-93-2
  • Molecular Weight : 150.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8264

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8264
Availability details for
Quantity
  • Availability
  • Add to Cart