COM448661579 Combi-Blocks, Inc.

Boc-lys(2-picolinoyl)-oh

  • MDL number MFCD03424220
  • Cas Number : 122532-80-3
  • Molecular Weight : 351.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8426
Availability details for
Quantity
  • Availability
  • Add to Cart