COM448661601 Combi-Blocks, Inc.

8-Bromoisoquinolin-3-amine

  • MDL number MFCD15526547
  • Cas Number : 1260760-06-2
  • Molecular Weight : 223.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8448

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8448
Availability details for
Quantity
  • Availability
  • Add to Cart