COM448661886 Combi-Blocks, Inc.

5-Bromo-3,4'-bipyridine

  • MDL number MFCD09908008
  • Cas Number : 106047-38-5
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8756

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8756
Availability details for
Quantity
  • Availability
  • Add to Cart