COM448661941 Combi-Blocks, Inc.

2-[2-(Trifluoromethyl)phenyl]ethan-1-amine

  • MDL number MFCD00128232
  • Cas Number : 774-18-5
  • Molecular Weight : 189.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8820

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8820
Availability details for
Quantity
  • Availability
  • Add to Cart