COM448661961 Combi-Blocks, Inc.

5-Chloro-2-methylnicotinaldehyde

  • MDL number MFCD10698573
  • Cas Number : 1060814-56-3
  • Molecular Weight : 155.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8841

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-8841
Availability details for
Quantity
  • Availability
  • Add to Cart