COM448662171 Combi-Blocks, Inc.

Cyclohexyl propionate

  • MDL number MFCD00046352
  • Cas Number : 6222-35-1
  • Molecular Weight : 156.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9071

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9071
Availability details for
Quantity
  • Availability
  • Add to Cart