COM448662195 Combi-Blocks, Inc.

Pentafluorophenyltriethoxysilane

  • MDL number MFCD03411257
  • Cas Number : 20083-34-5
  • Molecular Weight : 330.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9096
Availability details for
Quantity
  • Availability
  • Add to Cart