COM448662199 Combi-Blocks, Inc.

2-(3-Chloro-5-(trifluoromethyl)phenyl)acetic acid

  • MDL number MFCD04115851
  • Cas Number : 886496-99-7
  • Molecular Weight : 238.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9100
Availability details for
Quantity
  • Availability
  • Add to Cart