COM448662204 Combi-Blocks, Inc.

5-Phenyl-3h-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD00126441
  • Cas Number : 35978-39-3
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9105
Availability details for
Quantity
  • Availability
  • Add to Cart