COM448662217 Combi-Blocks, Inc.

3'-Hydroxy-4'-methylacetophenone

  • MDL number MFCD06658171
  • Cas Number : 33414-49-2
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9119
Availability details for
Quantity
  • Availability
  • Add to Cart