COM448662440 Combi-Blocks, Inc.

2-Ethoxyethanesulfonyl chloride

  • MDL number MFCD10032392
  • Cas Number : 69371-75-1
  • Molecular Weight : 172.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9362
Availability details for
Quantity
  • Availability
  • Add to Cart