COM448662456 Combi-Blocks, Inc.

2-Chloro-6,7-dihydro-5h-pyrrolo[3,4-d]pyrimidine HCl

  • MDL number MFCD17171437
  • Cas Number : 954232-71-4
  • Molecular Weight : 192.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9379

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9379
Availability details for
Quantity
  • Availability
  • Add to Cart