COM448662599 Combi-Blocks, Inc.

(1-Fluorovinyl)methyldiphenylsilane

  • MDL number MFCD08276379
  • Cas Number : 257610-49-4
  • Molecular Weight : 242.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9527

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9527
Availability details for
Quantity
  • Availability
  • Add to Cart