COM448662635 Combi-Blocks, Inc.

Boc-(s)-alpha-(4-chloro-benzyl)-proline

  • MDL number MFCD06796760
  • Cas Number : 1217777-96-2
  • Molecular Weight : 339.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9564

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9564
Availability details for
Quantity
  • Availability
  • Add to Cart