COM448662690 Combi-Blocks, Inc.

Chlorogenic acid hemihydrate

  • MDL number MFCD02179353
  • Cas Number : 6001-76-9
  • Molecular Weight : 726.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9619

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9619
Availability details for
Quantity
  • Availability
  • Add to Cart