COM448662961 Combi-Blocks, Inc.

2-Bromo-4-dimethoxymethyl-nitrobenzene

  • MDL number MFCD09878768
  • Cas Number : 929095-57-8
  • Molecular Weight : 276.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9906

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-9906
Availability details for
Quantity
  • Availability
  • Add to Cart