COM448663106 Combi-Blocks, Inc.

1'-Benzyl-6-chloro-7-fluoro-1h-spiro[furo[3,4-c]pyridine-3,4'-piperidin]-1-one

  • MDL number MFCD22573651
  • Cas Number : 1283090-74-3
  • Molecular Weight : 346.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0215

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0215
Availability details for
Quantity
  • Availability
  • Add to Cart