COM448663138 Combi-Blocks, Inc.

(S)-2-(Piperazin-2-yl)propan-2-ol 2HCl

  • MDL number MFCD23703246
  • Cas Number : 1319591-13-3
  • Molecular Weight : 217.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0344
Availability details for
Quantity
  • Availability
  • Add to Cart