COM448663287 Combi-Blocks, Inc.

(S)-4-Phenylpyrrolidin-2-one

  • MDL number MFCD22573804
  • Cas Number : 71657-88-0
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0890

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0890
Availability details for
Quantity
  • Availability
  • Add to Cart