COM448663462 Combi-Blocks, Inc.

Propargyl-peg4-t-butyl ester

  • MDL number MFCD22683285
  • Cas Number : 1355197-66-8
  • Molecular Weight : 316.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1542
Availability details for
Quantity
  • Availability
  • Add to Cart