COM448663499 Combi-Blocks, Inc.

Valeramide

  • MDL number MFCD00041895
  • Cas Number : 626-97-1
  • Molecular Weight : 101.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1684

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1684
Availability details for
Quantity
  • Availability
  • Add to Cart