COM448663531 Combi-Blocks, Inc.

2-((2-(2-(2-Methoxyethoxy)ethoxy)ethoxy)methyl)oxirane

  • MDL number MFCD22683314
  • Cas Number : 73692-54-3
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1805

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1805
Availability details for
Quantity
  • Availability
  • Add to Cart