COM448663567 Combi-Blocks, Inc.

Undecanophenone

  • MDL number MFCD00051548
  • Cas Number : 4433-30-1
  • Molecular Weight : 246.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1956

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1956
Availability details for
Quantity
  • Availability
  • Add to Cart