COM448663657 Combi-Blocks, Inc.

Allyl benzoate

  • MDL number MFCD00026104
  • Cas Number : 583-04-0
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2349

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2349
Availability details for
Quantity
  • Availability
  • Add to Cart