COM448663710 Combi-Blocks, Inc.

5,7-Diiodoimidazo[5,1-b]thiazole

  • MDL number MFCD23703183
  • Cas Number : 208722-50-3
  • Molecular Weight : 375.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2571
Availability details for
Quantity
  • Availability
  • Add to Cart