COM448663736 Combi-Blocks, Inc.

Hydroxy-PEG1-CH2CO2tBu

  • MDL number MFCD27635155
  • Cas Number : 287174-32-7
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2657
Availability details for
Quantity
  • Availability
  • Add to Cart