COM448663821 Combi-Blocks, Inc.

Ethyl 1-cyclohexyl-2-(furan-3-yl)-1h-benzo[d]imidazole-5-carboxylate

  • MDL number MFCD27991369
  • Cas Number : 871930-30-2
  • Molecular Weight : 338.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2940

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2940
Availability details for
Quantity
  • Availability
  • Add to Cart