COM448663823 Combi-Blocks, Inc.

(3-tert-Butoxycarbonylamino-oxetan-3-yl)acetic acid ethyl ester

  • MDL number MFCD22628756
  • Cas Number : 1416323-06-2
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2946
Availability details for
Quantity
  • Availability
  • Add to Cart