COM448663853 Combi-Blocks, Inc.

7-Amino-4-boc-4,5-dihydro-1h-benzo[e][1,4]diazepin-2(3h)-one

  • MDL number MFCD22377993
  • Cas Number : 1374651-40-7
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3054

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3054
Availability details for
Quantity
  • Availability
  • Add to Cart