COM448663875 Combi-Blocks, Inc.

7-(Bromomethyl)-2-chlorothieno[3,2-d]pyrimidine

  • MDL number MFCD23162762
  • Cas Number : 1356017-04-3
  • Molecular Weight : 263.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3135
Availability details for
Quantity
  • Availability
  • Add to Cart