COM448663997 Combi-Blocks, Inc.

3-(Trifluoroacetyl)phenylboronic acid, pinacol ester

  • MDL number MFCD18915048
  • Cas Number : 631909-42-7
  • Molecular Weight : 300.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3598
Availability details for
Quantity
  • Availability
  • Add to Cart