COM448664106 Combi-Blocks, Inc.

7-(tert-Butoxycarbonylamino)quinoline-3-carboxylic acid

  • MDL number MFCD22690563
  • Cas Number : 1416440-75-9
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4074

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4074
Availability details for
Quantity
  • Availability
  • Add to Cart