COM448664109 Combi-Blocks, Inc.

(-)-Alpha-[1-(dibutylamino)ethyl]benzyl alcohol

  • MDL number MFCD00142695
  • Cas Number : 114389-70-7
  • Molecular Weight : 263.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4082

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4082
Availability details for
Quantity
  • Availability
  • Add to Cart