COM448664110 Combi-Blocks, Inc.

5-Chloro-2-(methylthio)benzothiazole

  • MDL number MFCD00186100
  • Cas Number : 3507-41-3
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4083

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4083
Availability details for
Quantity
  • Availability
  • Add to Cart