COM448664147 Combi-Blocks, Inc.

tert-Butyl 1-methyl-2-oxopyrrolidin-3-ylcarbamate

  • MDL number MFCD22416370
  • Cas Number : 549531-10-4
  • Molecular Weight : 214.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4244

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4244
Availability details for
Quantity
  • Availability
  • Add to Cart