COM448664238 Combi-Blocks, Inc.

2-(4-Nitrophenyl)-1,3-thiazole

  • MDL number MFCD26794836
  • Cas Number : 3704-41-4
  • Molecular Weight : 206.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4615

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4615
Availability details for
Quantity
  • Availability
  • Add to Cart